Assay Details
| Gene Symbol: 4933407K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029443.1 | 2 - 3 | 331 | 64 | ||
| GenBank mRNA | AK016728.1 | - | 2 - 3 | 393 | 64 | |
| AK030939.1 | - | 2 - 3 | 332 | 64 | ||
| AK053637.1 | - | 2 - 3 | 282 | 64 | ||
| AK163097.1 | - | 331 | 64 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB39B, member RAS oncogene family |
Rab39b | 6330580M05Rik | -177.1kb | Sense | Chr. X - 75572045 - 75578231 on NCBI Build GRCm38 |
| Downstream | plastin 3 (T-isoform) | Pls3 |
AI115446, AL024105, T-fimbrin |
25.3kb | Sense | Chr. X - 75785654 - 75875170 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933407K13Rik
Genomic Map