Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029459.1 | 2 - 3 | 269 | 65 | ||
| GenBank mRNA | BC055849.1 | - | 2 - 3 | 189 | 65 | |
| CJ066501.1 | - | 2 - 3 | 207 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nucleolar RNA host gene 20 | Snhg20 | 2810008D09Rik | -38.2kb | Sense | Chr. 11 - 117076783 - 117078955 on NCBI Build GRCm38 |
| Downstream | septin 9 | Sept9 |
MSF1, Msf, PNUTL4, Sint1 |
70.9kb | Sense | Chr. 11 - 117199661 - 117362325 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l1
Genomic Map