Assay Details
| Gene Symbol: Mir154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029564.1 | 1 | 36 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 453 | Mir453 |
Mirn453, mmu-mir-453 |
-2.7kb | Sense | Chr. 12 - 109735619 - 109735700 on NCBI Build GRCm38 |
| Downstream | microRNA 496a | Mir496a |
Mir496, Mirn496, mir-496a, mmu-mir-496, mmu-mir-496a |
621.0 | Sense | Chr. 12 - 109739119 - 109739197 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir154
Genomic Map