Assay Details
| Gene Symbol: Mir182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029569.1 | 1 | 43 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
microRNA cluster 35, host gene |
Mirc35hg | Sense | Chr. 6 - 30158640 - 30174125 on NCBI Build GRCm38 | ||
| Downstream | microRNA 96 | Mir96 |
Mirn96, mir-96, mmu-mir-96 |
3.5kb | Sense | Chr. 6 - 30169446 - 30169551 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir182
Genomic Map