Assay Details
| Gene Symbol: Mir204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029591.1 | 1 | 41 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 41819 |
Gm41819 | -729.6kb | Sense | Chr. 19 - 21992200 - 22021023 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA C330002G04 gene | C330002G04Rik | 246.2kb | Anti-sense | Chr. 19 - 22996890 - 23075906 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir204
Genomic Map