Assay Details
| Gene Symbol: Mir19b-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029715.1 | 1 | 40 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 92-2 | Mir92-2 |
Mirn92-2, mir-92a-2 |
-55.0 | Sense | Chr. X - 52741838 - 52741928 on NCBI Build GRCm38 |
| Downstream | microRNA 20b | Mir20b |
Mirn20b, mir-20b, mmu-mir-20b |
47.0 | Sense | Chr. X - 52742113 - 52742192 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir19b-2
Genomic Map