Assay Details
| Gene Symbol: Mir30c-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029717.1 | 1 | 49 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 30a | Mir30a |
Mirn30a, mir-30a, mmu-mir-30a |
-19.4kb | Sense | Chr. 1 - 23272269 - 23272339 on NCBI Build GRCm38 |
| Downstream | opioid growth factor receptor-like 1 | Ogfrl1 | 2210417C17Rik | 74.6kb | Anti-sense | Chr. 1 - 23366424 - 23383210 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir30c-2
Genomic Map