Assay Details
| Gene Symbol: Mirlet7a-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029726.1 | 1 | 46 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 100 | Mir100 |
Mirn100, mir-100, mmu-mir-100 |
-5.2kb | Sense | Chr. 9 - 41531425 - 41531504 on NCBI Build GRCm38 |
| Downstream | microRNA 125b-1 | Mir125b-1 |
Mirn125b, Mirn125b-1, mir-125b-1 |
45.1kb | Sense | Chr. 9 - 41581926 - 41582002 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7a-2
Genomic Map