Assay Details
| Gene Symbol: Mir26a-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029742.1 | 1 | 46 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102636563 | LOC102636563 | -125.8kb | Anti-sense | Chr. 9 - 118902831 - 118906018 on NCBI Build GRCm38 | |
| Downstream | villin-like | Vill | Villp | 20.8kb | Sense | Chr. 9 - 119052729 - 119071525 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir26a-1
Genomic Map