Assay Details
| Gene Symbol: Mir29a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029744.1 | 1 | 41 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Kruppel-like factor 14 | Klf14 |
5330411L03Rik, AL022736, BTEB5 |
-103.7kb | Sense | Chr. 6 - 30956021 - 30958990 on NCBI Build GRCm38 |
| Downstream | microRNA 29b-1 | Mir29b-1 |
Mirn29b, Mirn29b-1, mir-29b-1 |
276.0 | Sense | Chr. 6 - 31063023 - 31063093 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir29a
Genomic Map