Assay Details
| Gene Symbol: Mir27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029746.1 | 1 | 47 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 23a | Mir23a |
Mirn23a, mir-23a, mmu-mir-23a |
-80.0 | Sense | Chr. 8 - 84208518 - 84208592 on NCBI Build GRCm38 |
| Downstream | microRNA 24-2 | Mir24-2 |
Mirn24-2, mir-24-2 |
57.0 | Sense | Chr. 8 - 84208815 - 84208921 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir27a
Genomic Map