Assay Details
| Gene Symbol: Mir92-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029748.1 | 1 | 35 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 363 | Mir363 |
Mirn363, mir-363, mmu-mir-363 |
-71.0 | Sense | Chr. X - 52741693 - 52741767 on NCBI Build GRCm38 |
| Downstream | microRNA 19b-2 | Mir19b-2 |
Mirn19b, Mirn19b-2, miR-19b, mir-19b-2 |
55.0 | Sense | Chr. X - 52741983 - 52742066 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir92-2
Genomic Map