Assay Details
| Gene Symbol: Mir96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029750.1 | 1 | 46 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
microRNA cluster 35, host gene |
Mirc35hg | Sense | Chr. 6 - 30158640 - 30174125 on NCBI Build GRCm38 | ||
| Downstream | microRNA 183 | Mir183 |
Mirn183, mir-183, mmu-mir-183 |
117.0 | Sense | Chr. 6 - 30169668 - 30169737 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir96
Genomic Map