Assay Details
| Gene Symbol: Mir212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029794.1 | 1 | 50 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hypermethylated in cancer 1 | Hic1 |
AA408311, HIC-1 |
-3.1kb | Anti-sense | Chr. 11 - 75164565 - 75170255 on NCBI Build GRCm38 |
| Downstream | microRNA 132 | Mir132 |
Mirn132, mir-132, mmu-mir-132 |
204.0 | Sense | Chr. 11 - 75173682 - 75173747 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir212
Genomic Map