Assay Details
| Gene Symbol: Mir33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029804.1 | 1 | 35 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterol regulatory element binding factor 2 | Srebf2 |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
Sense | Chr. 15 - 82147269 - 82205379 on NCBI Build GRCm38 | |
| Downstream | shisa family member 8 | Shisa8 | Orf26 | 8.8kb | Anti-sense | Chr. 15 - 82206952 - 82212815 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir33
Genomic Map