Assay Details
| Gene Symbol: Mir365-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029959.1 | 1 | 54 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 193a | Mir193a |
Mir193, Mirn193, mir-193a, mmu-mir-193, mmu-mir-193a |
-14.4kb | Sense | Chr. 11 - 79711969 - 79712034 on NCBI Build GRCm38 |
| Downstream |
UTP6, small subunit (SSU) processome component, homolog (yeast) |
Utp6 |
4732497O03Rik, HCA66 |
205.8kb | Anti-sense | Chr. 11 - 79932320 - 79962426 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir365-2
Genomic Map