Assay Details
| Gene Symbol: Mir494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030269.1 | 1 | 32 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 329 | Mir329 |
Mirn329, mir-329, mmu-mir-329 |
-1.7kb | Sense | Chr. 12 - 109713481 - 109713577 on NCBI Build GRCm38 |
| Downstream | microRNA 679 | Mir679 |
Mirn679, mir-679, mmu-mir-679 |
175.0 | Sense | Chr. 12 - 109715577 - 109715650 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir494
Genomic Map