Assay Details
| Gene Symbol: Mir487b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030271.1 | 1 | 35 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 381 | Mir381 |
Mirn381, mir-381, mmu-mir-381 |
-437.0 | Sense | Chr. 12 - 109726822 - 109726896 on NCBI Build GRCm38 |
| Downstream | microRNA 539 | Mir539 |
Mirn539, mir-539, mmu-mir-539 |
715.0 | Sense | Chr. 12 - 109728129 - 109728202 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir487b
Genomic Map