Assay Details
| Gene Symbol: Mir676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030525.1 | 1 | 47 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 28 | Tmem28 |
EG620592, Fam155b, Tnen28 |
-534.8kb | Sense | Chr. X - 99821069 - 99846345 on NCBI Build GRCm38 |
| Downstream | acyl-CoA wax alcohol acyltransferase 2 | Awat2 |
9430062J17Rik, ARAT, Dgat2l4, WS |
18.2kb | Anti-sense | Chr. X - 100399336 - 100494221 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir676
Genomic Map