Assay Details
| Gene Symbol: Mir105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030546.1 | 1 | 47 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 767 | Mir767 | mmu-mir-767 | -1.4kb | Sense | Chr. X - 72589988 - 72590094 on NCBI Build GRCm38 |
| Downstream |
gamma-aminobutyric acid (GABA) A receptor, subunit theta |
Gabrq | 233.6kb | Anti-sense | Chr. X - 72825163 - 72842607 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir105
Genomic Map