Assay Details
| Gene Symbol: Mir511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030609.1 | 1 | 31 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 236 | Tmem236 |
2010003H20Rik, Fam23a, Gm613 |
-39.0kb | Sense | Chr. 2 - 14174524 - 14221993 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 39 (zinc transporter), member 12 |
Slc39a12 |
AW046938, Gm731 |
127.2kb | Sense | Chr. 2 - 14388316 - 14494978 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir511
Genomic Map