Assay Details
| Gene Symbol: 1700001G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033199.1 | 1 | 752 | 58 | ||
| GenBank mRNA | AK008236.1 | - | 1 | 735 | 58 | |
| AK008325.1 | - | 1 | 735 | 58 | ||
| BC132564.1 | - | 1 | 555 | 58 | ||
| BC132566.1 | - | 1 | 555 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA primase, p58 subunit |
Prim2 | AI323589 | -30.0 | Anti-sense | Chr. 1 - 33453808 - 33669794 on NCBI Build GRCm38 |
| Downstream |
RAB23, member RAS oncogene family |
Rab23 |
AW545388, opb, opb2 |
49.2kb | Sense | Chr. 1 - 33719896 - 33742564 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700001G17Rik
Genomic Map