Assay Details
| Gene Symbol: 2900079G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033432.1 | 1 - 2 | 94 | 70 | ||
| GenBank mRNA | AK048592.1 | - | 1 - 2 | 78 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cyclic AMP-regulated phosphoprotein, 21 |
Arpp21 |
0710001E13Rik, AI853636, ARPP-21, D9Bwg1012e, R3hdm3, Tarpp |
Anti-sense | Chr. 9 - 112065091 - 112236022 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 36367 |
Gm36367 | 1.0mb | Sense | Chr. 9 - 113287712 - 113310985 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2900079G21Rik
Genomic Map