Assay Details
| Gene Symbol: Gm10466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033491.1 | 1 - 2 | 65 | 87 | ||
| GenBank mRNA | AK135806.1 | - | 1 - 2 | 66 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933430M04 gene | 4933430M04Rik | -156.2kb | Anti-sense | Chr. 11 - 24481909 - 24569951 on NCBI Build GRCm38 | |
| Downstream | predicted gene 12069 | Gm12069 | OTTMUSG00000005297 | 570.5kb | Anti-sense | Chr. 11 - 25301259 - 25322784 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135806.1 transcripts.
Set Membership: |
|

Gm10466
Genomic Map