Assay Details
| Gene Symbol: Gm10466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033491.1 | 2 - 3 | 158 | 94 | ||
| GenBank mRNA | AK135806.1 | - | 2 - 3 | 159 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933430M04 gene | 4933430M04Rik | -151.1kb | Anti-sense | Chr. 11 - 24481909 - 24569951 on NCBI Build GRCm38 | |
| Downstream | predicted gene 12069 | Gm12069 | OTTMUSG00000005297 | 575.0kb | Anti-sense | Chr. 11 - 25301259 - 25322784 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK135806.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098583 | NR_033491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098583 | AK135806.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09257863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09235214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09213832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09427377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09460759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09697572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09460754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08995959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09254876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09135755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09391632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08995953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08995946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09257856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09241115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09403687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09410630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09491703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09273146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09410635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09460755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27653086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27652993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.1 ST Array | 10384768 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100022465.mm.1 | NR_033491.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100022465.mm.1 | NR_033491.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100022465.mm.1 | TC1100002389.mm.1 | MTA TC |

Gm10466
Genomic Map