Assay Details
| Gene Symbol: Gm7271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033501.1 | 1 - 2 | 369 | 97 | ||
| GenBank mRNA | BC147734.1 | - | 1 - 2 | 452 | 9056683 | 97 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuromedin U | Nmu | -120.3kb | Anti-sense | Chr. 5 - 76333495 - 76363777 on NCBI Build GRCm38 | |
| Downstream | exocyst complex component 1 | Exoc1 |
2810407P21Rik, A730011E05Rik, SEC3, Sec3l1, Sec3p |
28.7kb | Sense | Chr. 5 - 76529311 - 76570298 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm7271
Genomic Map