Assay Details
| Gene Symbol: C030023E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033502.1 | 1 | 553 | 99 | ||
| GenBank mRNA | AK047748.1 | - | 1 | 553 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cerebellar degeneration related antigen 1 | Cdr1 |
100039759, Cdr, Cdr34, EG631990, Gm2409, Gm7077 |
-5.7kb | Anti-sense | Chr. X - 61183246 - 61185558 on NCBI Build GRCm38 |
| Downstream | predicted gene 4988 | Gm4988 | EG245440 | 96.9kb | Sense | Chr. X - 61291047 - 61292488 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030023E24Rik
Genomic Map