Assay Details
| Gene Symbol: Gm11110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033508.1 | 3 - 4 | 283 | 80 | ||
| GenBank mRNA | AK154177.1 | - | 3 - 4 | 282 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tubulin, beta 4A class IVA |
Tubb4a |
AI325297, M(beta)4, Tubb, Tubb4 |
-4.2kb | Sense | Chr. 17 - 57080066 - 57087782 on NCBI Build GRCm38 |
| Downstream |
tumor necrosis factor (ligand) superfamily, member 9 |
Tnfsf9 |
4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l |
3.1kb | Anti-sense | Chr. 17 - 57105277 - 57107757 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm11110
Genomic Map