Assay Details
| Gene Symbol: Gm10845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033535.1 | 1 - 2 | 156 | 89 | ||
| GenBank mRNA | AK143924.1 | - | 1 - 2 | 157 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protocadherin 8 | Pcdh8 |
1700080P15Rik, Papc |
-97.0kb | Sense | Chr. 14 - 79766772 - 79771312 on NCBI Build GRCm38 |
| Downstream | uncharacterized LOC328448 | LOC328448 | E130119H08 | 21.8kb | Anti-sense | Chr. 14 - 79890972 - 79984136 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143924.1 transcripts.
Set Membership: |
|

Gm10845
Genomic Map