Assay Details
| Gene Symbol: Gm7134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033597.1 | 1 | 2246 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6377 | Gm6377 | EG622976 | -1.2mb | Sense | Chr. X - 109191425 - 109200448 on NCBI Build GRCm38 |
| Downstream |
POU domain, class 3, transcription factor 4 |
Pou3f4 |
Brn-4, Brn4, OTF-9, Otf9, Slf, oct-9 |
439.3kb | Sense | Chr. X - 110814465 - 110815550 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm7134
Genomic Map