Assay Details
| Gene Symbol: Gm4349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033637.1 | 1 - 2 | 53 | 78 | ||
| GenBank mRNA | AV576091.1 | - | 1 - 2 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930558C23 gene | 4930558C23Rik | -24.5kb | Anti-sense | Chr. 3 - 95386458 - 95402841 on NCBI Build GRCm38 | |
| Downstream | aryl hydrocarbon receptor nuclear translocator | Arnt |
D3Ertd557e, Drnt, ESTM42, Hif1b, W08714, bHLHe2, mKIAA4051 |
5.5kb | Sense | Chr. 3 - 95434368 - 95497240 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4349
Genomic Map