Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 47 |
Snora47 | MBI-115 | Sense | Chr. 13 - 95330611 - 95330736 on NCBI Build GRCm38 | |
| Downstream | angiogenic factor with G patch and FHA domains 1 | Aggf1 |
2010009L17Rik, 2310029P06Rik, AW112072, Peg3, VG5Q |
19.9kb | Anti-sense | Chr. 13 - 95350683 - 95375357 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map