Assay Details
| Gene Symbol: Mir1964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035490.1 | 1 | 37 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38983 |
Gm38983 | -5.8kb | Anti-sense | Chr. 7 - 29763788 - 29767450 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 30 | Zfp30 |
2610306P15Rik, Zfp-30 |
11.4kb | Sense | Chr. 7 - 29784790 - 29794540 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir1964
Genomic Map