Assay Details
| Gene Symbol: Mir1966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035492.1 | 1 | 51 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | agouti related neuropeptide | Agrp |
Agrt, Art |
-35.6kb | Anti-sense | Chr. 8 - 105566695 - 105579845 on NCBI Build GRCm38 |
| Downstream | predicted gene 5914 | Gm5914 | 8.8kb | Anti-sense | Chr. 8 - 105624336 - 105637564 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir1966
Genomic Map