Assay Details
| Gene Symbol: Gm3086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036607.1 | 2 - 3 | 179 | 82 | ||
| GenBank mRNA | BC027570.1 | - | 2 - 3 | 115 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | salvador homolog 1 (Drosophila) | Sav1 |
1700040G09Rik, Salv, Sav, WW45, Wwp3, Wwp4 |
Anti-sense | Chr. 12 - 69965012 - 69987002 on NCBI Build GRCm38 | |
| Downstream | ninein | Nin |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
41.7kb | Anti-sense | Chr. 12 - 70011435 - 70113830 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027570.1 transcripts.
Set Membership: |
|

Gm3086
Genomic Map