Assay Details
| Gene Symbol: Mir30d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029718.1 | 1 | 43 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 30b | Mir30b |
Mirn30b, mir-30b, mmu-mir-30b |
-3.7kb | Sense | Chr. 15 - 68337415 - 68337510 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 41341 |
Gm41341 | 537.0kb | Anti-sense | Chr. 15 - 68878321 - 68883576 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir30d
Genomic Map