Assay Details
| Gene Symbol: Zfp389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026798.1 | 2 | 847 | 92 | ||
| GenBank mRNA | AK132930.1 | - | 2 | 848 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cytochrome c oxidase, subunit Vb pseudogene |
Gm11273 | OTTMUSG00000000426 | -2.9kb | Sense | Chr. 13 - 21500935 - 21501452 on NCBI Build GRCm38 |
| Downstream | zinc finger with KRAB and SCAN domains 8 | Zkscan8 |
2510038J07Rik, D430019P06Rik, LD5-1, Zfp192 |
8.0kb | Sense | Chr. 13 - 21513221 - 21531114 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp389
Genomic Map