Assay Details
| Gene Symbol: Josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205070.1 | NP_001191999.1 | 4 - 5 | 611 | 92 | |
| NM_001205071.1 | NP_001192000.1 | 3 - 4 | 499 | 92 | ||
| NM_001205072.1 | NP_001192001.1 | 3 - 4 | 468 | 92 | ||
| NM_001205073.1 | NP_001192002.1 | 3 - 4 | 465 | 92 | ||
| NM_025368.4 | NP_079644.1 | 3 - 4 | 496 | 92 | ||
| GenBank mRNA | AK002872.1 | - | 3 - 4 | 337 | 92 | |
| AK003525.1 | - | 3 - 4 | 421 | 92 | ||
| CA326008.1 | - | 3 - 4 | 410 | 92 | ||
More Information
Set Membership: |
|

Josd2
Genomic Map