Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026998.mm.1 |
NM_001195255.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026998.mm.1 |
NM_001195255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026998.mm.1 |
TC0700002995.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027001.mm.1 |
NM_001195255.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027001.mm.1 |
NM_001195255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027001.mm.1 |
TC0700002995.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027000.mm.1 |
NM_001195255.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027000.mm.1 |
NM_001195255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027000.mm.1 |
TC0700002995.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026999.mm.1 |
NM_001195255.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026999.mm.1 |
NM_001195255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026999.mm.1 |
TC0700002995.mm.1 |
MTA TC |