Assay Details
| Gene Symbol: Gfy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195255.1 | NP_001182184.1 | 4 - 5 | 1594 | 94 | |
| GenBank mRNA | AB845286.1 | - | 2 - 3 | 1321 | 94 | |
| BM934591.1 | - | 2 - 3 | 478 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gfy
Genomic Map