Assay Details
| Gene Symbol: Snord123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028575.2 | 1 | 45 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nucleolar RNA host gene 18 | Snhg18 |
0610007N19Rik, Snord123 |
Sense | Chr. 15 - 32240568 - 32244662 on NCBI Build GRCm38 | |
| Downstream |
sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A |
Sema5a |
5930434A13, 9130201M22Rik, AI464145, Semaf, semF |
2.8kb | Anti-sense | Chr. 15 - 32244742 - 32696341 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord123
Genomic Map