Assay Details
| Gene Symbol: 9330179D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040273.1 | 1 - 2 | 199 | 60 | ||
| GenBank mRNA | CX238860.1 | - | 1 - 2 | 72 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin D2 | Ccnd2 |
2600016F06Rik, AI256817, BF642806, C86853, Vin-1, Vin1, cD2 |
Anti-sense | Chr. 6 - 127125162 - 127152188 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 38404 |
Gm38404 | D130058E03 | 110.6kb | Anti-sense | Chr. 6 - 127296186 - 127307967 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9330179D12Rik
Genomic Map