Assay Details
| Gene Symbol: A730056A06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040324.1 | 1 - 2 | 260 | 101 | ||
| GenBank mRNA | BF470505.1 | - | 1 - 2 | 260 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100505025 | LOC100505025 | -17.2kb | Anti-sense | Chr. 7 - 73310380 - 73315193 on NCBI Build GRCm38 | |
| Downstream | chromodomain helicase DNA binding protein 2 | Chd2 |
2810013C04Rik, 2810040A01Rik, 5630401D06Rik, AI851092, BC029703, CHD-2 |
50.9kb | Sense | Chr. 7 - 73426652 - 73541746 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A730056A06Rik
Genomic Map