Assay Details
| Gene Symbol: Gm16023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040441.1 | 1 - 2 | 402 | 116 | ||
| GenBank mRNA | AK142359.1 | - | 1 - 2 | 403 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NAD kinase | Nadk |
4432404C02Rik, BC004012 |
-19.4kb | Anti-sense | Chr. 4 - 155562392 - 155591001 on NCBI Build GRCm38 |
| Downstream | cyclin-dependent kinase 11B | Cdk11b |
AA989746, CDK11-p110, CDK11-p46, CDK11-p58, Cdc11b, Cdc2l1, Cdc2l2, Cdk11, p58 |
91.0 | Anti-sense | Chr. 4 - 155624846 - 155649935 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16023
Genomic Map