Assay Details
| Gene Symbol: Gm5441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_044987.1 | 1 - 2 | 128 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, receptor type, N polypeptide 2 |
Ptprn2 |
4930425H11Rik, IA2beta, PTP-NP, Phol, mKIAA0387, phogrin |
Anti-sense | Chr. 12 - 116485692 - 117278167 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 39559 |
Gm39559 | 8.6kb | Anti-sense | Chr. 12 - 117345562 - 117347995 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5441
Genomic Map