Assay Details
| Gene Symbol: Gdap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045032.1 | 1 | 1820 | 106 | ||
| GenBank mRNA | BC052902.1 | - | 1 | 1820 | 30067675 | 106 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nicotinamide phosphoribosyltransferase | Nampt |
1110035O14Rik, AI314458, AI480535, NAmPRTase, Pbef, Pbef1, Visfatin |
Sense | Chr. 12 - 32820335 - 32853369 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4933406C10 gene | 4933406C10Rik | 92.5kb | Anti-sense | Chr. 12 - 32919384 - 32953789 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdap10
Genomic Map