Assay Details
| Gene Symbol: Gm10336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045170.1 | 1 | 2922 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 5-methyltetrahydrofolate-homocysteine methyltransferase | Mtr |
AI894170, D830038K18Rik, MS |
Sense | Chr. 13 - 12182720 - 12258195 on NCBI Build GRCm38 | |
| Downstream | actinin alpha 2 | Actn2 | 1110008F24Rik | 82.9kb | Sense | Chr. 13 - 12269426 - 12340732 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm10336
Genomic Map