Assay Details
| Gene Symbol: 2900055J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045177.1 | 1 | 519 | 107 | ||
| GenBank mRNA | AK013700.1 | - | 1 | 520 | 107 | |
| BM932606.1 | - | 1 | 441 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31087 |
Gm31087 | -10.0kb | Anti-sense | Chr. 18 - 40242893 - 40246936 on NCBI Build GRCm38 | |
| Downstream | potassium channel tetramerisation domain containing 16 | Kctd16 |
4930434H12Rik, Gm1267 |
658.0 | Sense | Chr. 18 - 40258361 - 40531184 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900055J20Rik
Genomic Map