Assay Details
| Gene Symbol: 1110015O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045272.1 | 3 - 4 | 329 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
hepatocyte nuclear factor 4, gamma |
Hnf4g | NR2A2 | -1.1mb | Sense | Chr. 3 - 3508030 - 3659800 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 17308 |
Gm17308 | 275.4kb | Sense | Chr. 3 - 5090276 - 5093402 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1110015O18Rik
Genomic Map