Assay Details
| Gene Symbol: 1700074H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045296.1 | 2 - 3 | 283 | 87 | ||
| GenBank mRNA | AK006947.1 | - | 2 - 3 | 294 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chemokine (C-C motif) ligand 28 | Ccl28 |
CCK1, MEC, Scya28 |
-26.4kb | Sense | Chr. 13 - 119610551 - 119654359 on NCBI Build GRCm38 |
| Downstream | 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 | Hmgcs1 | B130032C06Rik | 8.8kb | Sense | Chr. 13 - 119690351 - 119708260 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700074H08Rik
Genomic Map